Assay Details
Gene Symbol: LOC4324047 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015760852.1 | XP_015616338.1 | 2 - 3 | 594 | 70 | |
XM_015760857.2 | XP_015616343.1 | 2 - 3 | 556 | 70 | ||
GenBank mRNA | AK059119.1 | - | 2 - 3 | 562 | 70 | |
AK102891.1 | - | 2 - 3 | 562 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
