Assay Details
Gene Symbol: LOC4324047 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015760857.2 | XP_015616343.1 | 3 - 4 | 597 | 69 | |
GenBank mRNA | AK059119.1 | - | 3 - 4 | 603 | 69 | |
AK102891.1 | - | 3 - 4 | 603 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
