Assay Details
Gene Symbol: LOC4324560 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788170.2 | XP_015643656.1 | 1 - 2 | 567 | 58 | |
GenBank mRNA | AK111255.1 | - | 1 - 2 | 572 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
