Assay Details
| Gene Symbol: LOC4327665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756464.2 | XP_015611950.1 | 2 - 3 | 282 | 97 | |
| GenBank mRNA | AJ251563.1 | - | 2 - 3 | 178 | 97 | |
| AK070745.1 | - | 2 - 3 | 238 | 97 | ||
| AK104419.1 | - | 2 - 3 | 183 | 97 | ||
More Information
Set Membership: |
|

LOC4327665
Genomic Map