Assay Details
| Gene Symbol: LOC4327665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756464.2 | XP_015611950.1 | 4 - 5 | 493 | 97 | |
| GenBank mRNA | AJ251563.1 | - | 4 - 5 | 389 | 97 | |
| AK070745.1 | - | 4 - 5 | 449 | 97 | ||
| AK104055.1 | - | 4 - 5 | 405 | 97 | ||
| AK104419.1 | - | 4 - 5 | 394 | 97 | ||
More Information
Set Membership: |
|

LOC4327665
Genomic Map