Assay Details
| Gene Symbol: LOC4327862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759452.2 | XP_015614938.1 | 2 - 3 | 230 | 106 | |
| GenBank mRNA | AK058452.1 | - | 2 - 3 | 236 | 106 | |
| AK060072.1 | - | 2 - 3 | 224 | 106 | ||
More Information
Set Membership: |
|

LOC4327862
Genomic Map