Assay Details
Gene Symbol: LOC4324725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794859.2 | XP_015650345.1 | 1 - 2 | 381 | 72 | |
GenBank mRNA | AK073926.1 | - | 1 - 2 | 377 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
