Assay Details
Gene Symbol: LOC4324725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794859.2 | XP_015650345.1 | 2 - 3 | 515 | 77 | |
GenBank mRNA | AK073926.1 | - | 2 - 3 | 511 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
