Assay Details
Gene Symbol: LOC4324727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794815.2 | XP_015650301.1 | 1 - 2 | 613 | 67 | |
GenBank mRNA | AK063441.1 | - | 1 - 2 | 682 | 67 | |
AK101974.1 | - | 1 - 2 | 1137 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
