Assay Details
Gene Symbol: LOC4324727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794815.2 | XP_015650301.1 | 3 - 4 | 1715 | 94 | |
GenBank mRNA | AK101974.1 | - | 3 - 4 | 2239 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
