Assay Details
Gene Symbol: LOC4324727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794815.2 | XP_015650301.1 | 5 - 6 | 2173 | 110 | |
GenBank mRNA | AK101974.1 | - | 4 - 5 | 2825 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
