Assay Details
| Gene Symbol: LOC4327034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK061563.1 | - | 1 - 2 | 161 | 95 | |
| AK066596.1 | - | 1 - 2 | 205 | 95 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4327034| Gene Symbol: LOC4327034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK061563.1 | - | 1 - 2 | 161 | 95 | |
| AK066596.1 | - | 1 - 2 | 205 | 95 | ||
Set Membership: |
|