Assay Details
| Gene Symbol: LOC4327034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK061563.1 | - | 3 - 4 | 517 | 92 | |
| AK066596.1 | - | 3 - 4 | 561 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4327034| Gene Symbol: LOC4327034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK061563.1 | - | 3 - 4 | 517 | 92 | |
| AK066596.1 | - | 3 - 4 | 561 | 92 | ||
Set Membership: |
|