Assay Details
Gene Symbol: LOC4325503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765411.1 | XP_015620897.1 | 1 - 2 | 1187 | 100 | |
GenBank mRNA | AK067198.1 | - | 1 - 2 | 1186 | 100 | |
DQ450949.1 | - | 1 - 2 | 729 | 100 | ||
HM991166.1 | - | 1 - 2 | 704 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
