Assay Details
Gene Symbol: LOC4325503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765411.1 | XP_015620897.1 | 3 - 4 | 2132 | 68 | |
GenBank mRNA | AK067198.1 | - | 3 - 4 | 2131 | 68 | |
BT129328.1 | - | 2 - 3 | 349 | 68 | ||
DQ450949.1 | - | 3 - 4 | 1674 | 68 | ||
HM991166.1 | - | 3 - 4 | 1649 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
