Assay Details
Gene Symbol: LOC4324711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765311.2 | XP_015620797.1 | 11 - 12 | 1266 | 76 | |
XM_015765321.2 | 7 - 8 | 521 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
