Assay Details
Gene Symbol: LOC4324711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765311.2 | XP_015620797.1 | 18 - 19 | 1883 | 145 | |
XM_015765321.2 | 14 - 15 | 1138 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() |
