Assay Details
Gene Symbol: LOC4324711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765311.2 | XP_015620797.1 | 26 - 27 | 2663 | 111 | |
XM_015765321.2 | 22 - 23 | 1918 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
