Assay Details
Gene Symbol: LOC4324711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765311.2 | XP_015620797.1 | 33 - 34 | 3348 | 90 | |
XM_015765321.2 | 29 - 30 | 2603 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
