Assay Details
Gene Symbol: LOC4324711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765311.2 | XP_015620797.1 | 42 - 43 | 4774 | 98 | |
XM_015765321.2 | 38 - 39 | 4029 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
