Assay Details
| Gene Symbol: LOC4324952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765634.2 | XP_015621120.1 | 5 - 6 | 862 | 101 | |
| XM_015765635.1 | XP_015621121.1 | 5 - 6 | 696 | 101 | ||
| GenBank mRNA | AK066633.1 | - | 5 - 6 | 902 | 101 | |
| AK100381.1 | - | 5 - 6 | 859 | 101 | ||
More Information
Set Membership: |
|

LOC4324952
Genomic Map