Assay Details
Gene Symbol: LOC4328006 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771364.2 | XP_015626850.1 | 12 - 13 | 1461 | 101 | |
GenBank mRNA | AK288268.1 | - | 8 - 9 | 2584 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
