Assay Details
Gene Symbol: LOC4328006 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771364.2 | XP_015626850.1 | 2 - 3 | 641 | 69 | |
GenBank mRNA | AK288268.1 | - | 2 - 3 | 633 | 69 | |
DQ539442.1 | - | 1 - 2 | 143 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
