Assay Details
Gene Symbol: LOC4328006 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771364.2 | XP_015626850.1 | 4 - 5 | 806 | 75 | |
GenBank mRNA | AK288268.1 | - | 4 - 5 | 798 | 75 | |
DQ539442.1 | - | 3 - 4 | 308 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
