Assay Details
Gene Symbol: LOC4328006 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771364.2 | XP_015626850.1 | 5 - 6 | 882 | 91 | |
GenBank mRNA | DQ539442.1 | - | 4 - 5 | 384 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
