Assay Details
Gene Symbol: LOC4328147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770212.2 | XP_015625698.1 | 2 - 3 | 305 | 133 | |
GenBank mRNA | AK101889.1 | - | 2 - 3 | 256 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
