Assay Details
Gene Symbol: LOC4328147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770212.2 | XP_015625698.1 | 3 - 4 | 624 | 123 | |
GenBank mRNA | AK101889.1 | - | 3 - 4 | 575 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
