Assay Details
Gene Symbol: LOC4328147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770212.2 | XP_015625698.1 | 9 - 10 | 1110 | 74 | |
GenBank mRNA | AK101889.1 | - | 9 - 10 | 1061 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
