Assay Details
Gene Symbol: LOC4346173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795284.2 | XP_015650770.1 | 1 - 2 | 943 | 110 | |
XM_015795285.2 | XP_015650771.1 | 1 - 2 | 944 | 110 | ||
GenBank mRNA | AK111766.1 | - | 1 - 2 | 715 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
