Assay Details
Gene Symbol: LOC4346173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795285.2 | XP_015650771.1 | 3 - 4 | 1130 | 94 | |
GenBank mRNA | AK111766.1 | - | 3 - 4 | 901 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
