Assay Details
Gene Symbol: LOC4346173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795284.2 | XP_015650770.1 | 9 - 10 | 1892 | 86 | |
XM_015795285.2 | XP_015650771.1 | 9 - 10 | 1890 | 86 | ||
GenBank mRNA | AK111766.1 | - | 9 - 10 | 1661 | 86 | |
AK112001.1 | - | 9 - 10 | 1357 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
