Assay Details
Gene Symbol: LOC4346180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795164.2 | XP_015650650.1 | 3 - 4 | 514 | 87 | |
GenBank mRNA | AK067119.1 | - | 3 - 4 | 499 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
