Assay Details
Gene Symbol: LOC4346180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795164.2 | XP_015650650.1 | 4 - 5 | 539 | 84 | |
GenBank mRNA | AK067119.1 | - | 4 - 5 | 524 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
