Assay Details
Gene Symbol: LOC4346188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015793018.2 | XP_015648504.1 | 1 - 2 | 309 | 67 | |
GenBank mRNA | AK106511.1 | - | 1 - 2 | 313 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
