Assay Details
Gene Symbol: LOC4346188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015793018.2 | XP_015648504.1 | 4 - 5 | 732 | 74 | |
GenBank mRNA | AK106511.1 | - | 4 - 5 | 736 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
