Assay Details
Gene Symbol: LOC4346188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015793018.2 | XP_015648504.1 | 7 - 8 | 960 | 109 | |
GenBank mRNA | AK106511.1 | - | 7 - 8 | 964 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
