Assay Details
Gene Symbol: LOC4346211 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794438.2 | XP_015649924.1 | 19 - 20 | 2371 | 126 | |
GenBank mRNA | AK071354.1 | - | 19 - 20 | 2261 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
