Assay Details
Gene Symbol: LOC4347053 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755999.2 | XP_015611485.1 | 1 | 47 | 72 | |
GenBank mRNA | AK058840.1 | - | 1 | 53 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015755999.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
