Assay Details
| Gene Symbol: LOC4352553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765060.1 | XP_015620546.1 | 1 - 2 | 417 | 66 | |
| GenBank mRNA | AK060201.1 | - | 1 - 2 | 417 | 66 | |
| AK070277.1 | - | 1 - 2 | 343 | 66 | ||
| AK071697.1 | - | 1 - 2 | 343 | 66 | ||
More Information
Set Membership: |
|

LOC4352553
Genomic Map