Assay Details
| Gene Symbol: LOC4352553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765060.1 | XP_015620546.1 | 2 - 3 | 544 | 79 | |
| GenBank mRNA | AK059954.1 | - | 3 - 4 | 235 | 79 | |
| AK060201.1 | - | 2 - 3 | 544 | 79 | ||
| AK070277.1 | - | 2 - 3 | 470 | 79 | ||
| AK071697.1 | - | 2 - 3 | 470 | 79 | ||
More Information
Set Membership: |
|

LOC4352553
Genomic Map