Assay Details
Gene Symbol: LOC4352706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764575.1 | XP_015620061.1 | 2 - 3 | 352 | 72 | |
GenBank mRNA | AK101904.1 | - | 2 - 3 | 359 | 72 | |
AK103799.1 | - | 2 - 3 | 124 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
