Assay Details
Gene Symbol: LOC4352706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764575.1 | XP_015620061.1 | 4 - 5 | 557 | 96 | |
GenBank mRNA | AK101904.1 | - | 4 - 5 | 564 | 96 | |
AK103799.1 | - | 4 - 5 | 329 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
