Assay Details
Gene Symbol: LOC4352706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764575.1 | XP_015620061.1 | 6 - 7 | 853 | 63 | |
GenBank mRNA | AK103799.1 | - | 6 - 7 | 625 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
