Assay Details
Gene Symbol: LOC4350543 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761205.2 | XP_015616691.1 | 1 - 2 | 41 | 94 | |
GenBank mRNA | AK067664.1 | - | 1 - 2 | 42 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015761205.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
