Assay Details
Gene Symbol: LOC4350543 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761205.2 | XP_015616691.1 | 2 - 3 | 107 | 123 | |
GenBank mRNA | AK067664.1 | - | 2 - 3 | 108 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
