Assay Details
Gene Symbol: LOC4350543 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015761205.2 | XP_015616691.1 | 4 - 5 | 1256 | 75 | |
GenBank mRNA | AK067664.1 | - | 4 - 5 | 1257 | 75 | |
AK104957.1 | - | 3 - 4 | 1180 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
