Assay Details
| Gene Symbol: LOC4351017 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759728.2 | XP_015615214.1 | 5 - 6 | 1098 | 134 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4351017| Gene Symbol: LOC4351017 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759728.2 | XP_015615214.1 | 5 - 6 | 1098 | 134 | |
Set Membership: |
|