Assay Details
| Gene Symbol: LOC4337574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784553.2 | XP_015640039.1 | 14 - 15 | 2757 | 79 | |
| GenBank mRNA | AK099809.1 | - | 7 - 8 | 1112 | 79 | |
| AK120821.1 | - | 11 - 12 | 1953 | 79 | ||
More Information
Set Membership: |
|

LOC4337574
Genomic Map