Assay Details
| Gene Symbol: LOC4337574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784553.2 | XP_015640039.1 | 23 - 24 | 4043 | 99 | |
| GenBank mRNA | AK099809.1 | - | 16 - 17 | 2398 | 99 | |
| AK120821.1 | - | 20 - 21 | 3238 | 99 | ||
More Information
Set Membership: |
|

LOC4337574
Genomic Map