Assay Details
| Gene Symbol: LOC4337574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784553.2 | XP_015640039.1 | 24 - 25 | 4093 | 102 | |
| GenBank mRNA | AK099809.1 | - | 17 - 18 | 2448 | 102 | |
| AK120821.1 | - | 21 - 22 | 3288 | 102 | ||
More Information
Set Membership: |
|

LOC4337574
Genomic Map